Undec-10-enoic acid 99.5% stabilised

Supplier: Ambeed
Warning

Synonyms: Undecylenic-10 acid, Undecenoic acid, 10-Undecenoic acid, Undecylenic acid, Undecenylic acid

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A997538-100G A997538-10G A997538-25G A997538-500G A997538-1000G
77007-300EA 20 USD
77007-300 77007-302 77007-304 77007-306 77674-444
Undec-10-enoic acid 99.5% stabilised
Undec-10-enoic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₀O₂
MDL Number: MFCD00004442
CAS Number: 112-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR