7-Bromo-6-nitro-1,2,3,4-tetrahydroisoquinoline 95%

Supplier: Ambeed
Warning

A996150-100MG A996150-250MG A996150-1G
77020-234EA 30.77 USD
77020-234 77020-236 77614-718
7-Bromo-6-nitro-1,2,3,4-tetrahydroisoquinoline 95%
7-Bromo-6-nitro-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O₂
MW: 257.08 g/mol
MDL Number: MFCD20483534
CAS Number: 186390-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR