3-(Pyrimidin-5-yl)prop-2-yn-1-ol 95%

Supplier: Ambeed
Warning

A991311-1G A991311-250MG
77020-230EA 102.91 USD
77020-230 77020-232
3-(Pyrimidin-5-yl)prop-2-yn-1-ol 95%
3-(Pyrimidin-5-yl)prop-2-yn-1-ol

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.13 g/mol
MDL Number: MFCD07022248
CAS Number: 174456-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR