(5-Fluoro-2-methyl-1H-indol-3-yl)acetic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Fluoro-2-methyl-1H-indol-3-yl)acetic acid

A990568-100MG A990568-1G A990568-250MG
77037-078EA 18.04 USD
77037-078 77037-080 77037-082
(5-Fluoro-2-methyl-1H-indol-3-yl)acetic acid ≥95%
(5-Fluoro-2-methyl-1H-indol-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀FNO₂
MW: 207.204 g/mol
MDL Number: MFCD02664389
CAS Number: 71987-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR