5-Chloro-2-(chloromethyl)pyridine hydrochloride 96%

Supplier: Ambeed
Danger

A990074-100MG A990074-1G A990074-5G
77020-226EA 20 USD
77020-226 77020-228 77891-740
5-Chloro-2-(chloromethyl)pyridine hydrochloride 96%
5-Chloro-2-(chloromethyl)pyridine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N·ClH
MDL Number: MFCD20921510
CAS Number: 124425-84-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR