3-Amino-4-fluorobenzylalkohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Amino-4-fluorophenyl)methanol

A989686-100MG A989686-1G A989686-250MG
77007-266EA 24 USD
77007-266 77007-268 77007-270
3-Amino-4-fluorobenzylalkohol 98%
3-Amino-4-fluorobenzylalkohol

Precaución: for Research Use Only

Formula: C₇H₈FNO
MDL Number: MFCD09865016
CAS Number: 227609-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR