(S,E)-N-(2-((tert-Butyldimethylsilyl)oxy)ethylidene)-2-methylpropane-2-sulfinamide 95%

Supplier: Ambeed
Warning

A987622-1G A987622-250MG
76994-092EA 614.27 USD
76994-092 76994-094
(S,E)-N-(2-((tert-Butyldimethylsilyl)oxy)ethylidene)-2-methylpropane-2-sulfinamide 95%
(S,E)-N-(2-((tert-Butyldimethylsilyl)oxy)ethylidene)-2-methylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₁₂H₂₇NO₂SSi
MW: 277.49 g/mol
MDL Number: MFCD18252944
CAS Number: 918413-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR