3-Chloro-4-phenoxyaniline 95%
Supplier: Ambeed
|
Warning
|
A987494-1G
A987494-250MG
77037-066EA
88.06
USD
77037-066
77037-068
3-Chloro-4-phenoxyaniline 95%
3-Chloro-4-phenoxyaniline
Precaución: for Research Use Only
Formula:
C₁₂H₁₀ClNO MW: 219.66 g/mol |
MDL Number:
MFCD00459647 CAS Number: 5335-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...