2-Methyl-1,3-oxazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxy-2-methyl-1,3-oxazole, 2-Methyloxazole-4-carboxylic acid

A987150-100G A987150-10G A987150-1G A987150-250MG A987150-25G A987150-5G
77020-208EA 2087.86 USD
77020-208 77020-210 77020-212 77020-214 77020-216 77020-218
2-Methyl-1,3-oxazole-4-carboxylic acid 98%
2-Methyl-1,3-oxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₃
MW: 127.098 g/mol
MDL Number: MFCD00234078
CAS Number: 23012-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR