2-Methyl-3-hydroxy-6-iodopyridine 95+%

Supplier: Ambeed
Warning

Synonyms: 6-Iodo-2-methylpyridin-3-ol

A986738-100MG A986738-1G
77020-204EA 33.95 USD
77020-204 77020-206
2-Methyl-3-hydroxy-6-iodopyridine 95+%
2-Methyl-3-hydroxy-6-iodopyridine

Precaución: for Research Use Only

Formula: C₆H₆INO
MW: 235.022 g/mol
MDL Number: MFCD11044376
CAS Number: 848952-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR