2-Nitro-4-thiophenecarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Nitrothiophene-3-carboxylic acid
A985957-1G
A985957-5G
A985957-25G
A985957-250MG
76994-088EA
29.71
USD
76994-088
76994-090
77614-708
77684-916
2-Nitro-4-thiophenecarboxylic acid 98%
2-Nitro-4-thiophenecarboxylic acid
Precaución: For research use only.
Formula:
C₅H₃NO₄S MW: 173.15 g/mol |
MDL Number:
MFCD00052582 CAS Number: 40357-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...