Quinoline-8-sulfonamide 95%

Supplier: Ambeed
Warning

A985791-100MG A985791-250MG
77037-062EA 80.63 USD
77037-062 77037-064
Quinoline-8-sulfonamide 95%
Quinoline-8-sulfonamide

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂S
MDL Number: MFCD01191906
CAS Number: 35203-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR