3,3-Dimethyl-2-pyrrolidinone 98%

Supplier: Ambeed
Warning

Synonyms: 3,3-Dimethylpyrrolidin-2-one

A985683-100MG A985683-250MG
76994-086EA 35.01 USD
76994-086 77573-944
3,3-Dimethyl-2-pyrrolidinone 98%
3,3-Dimethyl-2-pyrrolidinone

Precaución: for Research Use Only

Formula: C₆H₁₁NO
MW: 113.16 g/mol
MDL Number: MFCD16622813
CAS Number: 4831-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR