1-Acetylpiperidin-4-amine 96%

Supplier: Ambeed
Warning

Synonyms: 1-Acetyl-4-aminopiperidine

A984171-1G A984171-250MG A984171-25G A984171-5G
77007-250EA 29.71 USD
77007-250 77007-252 77007-254 77007-256
1-Acetylpiperidin-4-amine 96%
1-Acetylpiperidin-4-amine

Precaución: for Research Use Only

Formula: C₇H₁₄N₂O
MW: 142.2000 g/mol
MDL Number: MFCD03412449
CAS Number: 160357-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR