Benserazide hydrochloride 98%

Supplier: Ambeed
Warning

A983665-1G A983665-1MG A983665-250MG A983665-5G A983665-25G A983665-100G
77020-192EA 16 USD
77020-192 77020-194 77020-196 77614-702 77614-700 77893-778
Benserazide hydrochloride 98%
Benserazide hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₅N₃O₅
MDL Number: MFCD00078571
CAS Number: 14919-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR