Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
P(t-Bu)3 Pd G2
A983523-1G
A983523-250MG
A983523-5G
A983523-25G
77020-186EA
66.84
USD
77020-186
77020-188
77020-190
77684-913
Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II) 98%
Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II)
Precaución: For research use only.
Formula:
C₂₄H₃₇ClNPPd MW: 512.41 g/mol |
MDL Number:
MFCD21608496 CAS Number: 1375325-71-5 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...