1-(2-Hydroxyethyl)pyrrolidine 98%

Supplier: Ambeed
Danger

Synonyms: 1-Pyrrolidineethanol, N-(2-Hydroxyethyl)pyrrolidine

A983226-10G A983226-25G A983226-5G
77007-238EA 14.86 USD
77007-238 77007-240 77007-242
1-(2-Hydroxyethyl)pyrrolidine 98%
1-(2-Hydroxyethyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD00003181
CAS Number: 2955-88-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR