Methyl-5-morpholino-2-nitrobenzenecarboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-morpholino-2-nitrobenzoate
A983029-100MG
A983029-1G
A983029-250MG
77007-232EA
25.47
USD
77007-232
77007-234
77007-236
Methyl-5-morpholino-2-nitrobenzenecarboxylate 95%
Methyl-5-morpholino-2-nitrobenzenecarboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₄N₂O₅ MW: 266.26 g/mol |
MDL Number:
MFCD11841050 CAS Number: 134050-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...