2,5-Diaminopyridine 98%

Supplier: Ambeed
Warning

A981354-250MG A981354-5G A981354-1G
77020-174EA 16 USD
77020-174 77020-176 77891-735
2,5-Diaminopyridine 98%
2,5-Diaminopyridine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MDL Number: MFCD00129023
CAS Number: 4318-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR