Imidazo[1,2-a]pyrimidine-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A981195-100MG A981195-1G A981195-250MG A981195-5G
77020-170EA 21 USD
77020-170 77020-172 77891-733 77891-734
Imidazo[1,2-a]pyrimidine-3-carbaldehyde 98%
Imidazo[1,2-a]pyrimidine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₅N₃O
MDL Number: MFCD09027563
CAS Number: 106012-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR