3-Cyanobenzamide 97%

Supplier: Ambeed
Warning

A980914-1G A980914-250MG A980914-5G
77020-164EA 91.24 USD
77020-164 77020-166 77020-168
3-Cyanobenzamide 97%
3-Cyanobenzamide

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MDL Number: MFCD00017574
CAS Number: 3441-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR