3-Chloro-2-methoxyphenol 98%

Supplier: Ambeed
Warning

A980897-100MG A980897-5G
77020-160EA 72.15 USD
77020-160 77020-162
3-Chloro-2-methoxyphenol 98%
3-Chloro-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇ClO₂
MW: 158.58 g/mol
MDL Number: MFCD16998708
CAS Number: 77102-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR