2-Amino-6-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A978685-1G A978685-250MG
76994-082EA 37 USD
76994-082 76994-084
2-Amino-6-(trifluoromethyl)benzonitrile 98%
2-Amino-6-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂
MDL Number: MFCD08741381
CAS Number: 58458-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR