Cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Danger

A978561-100G A978561-10G A978561-25G A978561-500G
77007-224EA 9.55 USD
77007-224 77007-226 77007-228 77007-230
Cyclopropanecarboxylic acid 97%
Cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₄H₆O₂
MW: 86.09 g/mol
MDL Number: MFCD00001287
CAS Number: 1759-53-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR