tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture) 97%

Supplier: Ambeed
Warning

Synonyms: 5-Norbornene-2-carboxylic Acid tert-Butyl Ester, tert-Butyl Bicyclo[2.2.1]hept-5-ene-2-carboxylate, Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid tert-Butyl Ester, tert-Butyl Bicyclo[2.2.1]-5-heptene-2-carboxylate

A978456-100G A978456-10G A978456-25G
77007-218EA 50 USD
77007-218 77007-220 77007-222
tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture) 97%
tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture)

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₂
MDL Number: MFCD18083704
CAS Number: 154970-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR