3-(2-Chlorophenoxy)propionic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Chlorophenoxy)propanoic acid

A978376-100MG A978376-1G A978376-250MG
77007-212EA 33.95 USD
77007-212 77007-214 77007-216
3-(2-Chlorophenoxy)propionic acid 95%
3-(2-Chlorophenoxy)propionic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.6190 g/mol
MDL Number: MFCD02295728
CAS Number: 7170-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR