(S)-2-Methylazetidine hydrochloride 98%

Supplier: Ambeed
Warning

A977971-1G A977971-250MG
76994-080EA 152 USD
76994-080 77891-731
(S)-2-Methylazetidine hydrochloride 98%
(S)-2-Methylazetidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClN
MDL Number: MFCD29920004
CAS Number: 935669-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR