1-Bromo-4-(methylsulfonyl)-2-nitrobenzene 98%

Supplier: Ambeed
Danger

A976508-1G A976508-250MG A976508-5G
76994-074EA 47.75 USD
76994-074 76994-076 76994-078
1-Bromo-4-(methylsulfonyl)-2-nitrobenzene 98%
1-Bromo-4-(methylsulphonyl)-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₆BrNO₄S
MW: 280.0958 g/mol
MDL Number: MFCD00054605
CAS Number: 94832-06-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR