4-Isobutylaniline 97%

Supplier: Ambeed
Warning

A974924-1G A974924-250MG A974924-5G
77037-050EA 125 USD
77037-050 77037-052 77037-054
4-Isobutylaniline 97%
4-Isobutylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₅N
MDL Number: MFCD00130035
CAS Number: 30090-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR