2-Bromo-5-(trifluoromethoxy)benzaldehyde 96%

Supplier: Ambeed
Warning

A973659-1G A973659-250MG A973659-100MG
76994-070EA 100 USD
76994-070 76994-072 77891-729
2-Bromo-5-(trifluoromethoxy)benzaldehyde 96%
2-Bromo-5-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
MDL Number: MFCD07368722
CAS Number: 505084-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR