4,4-dinitro-2,2-Bipyridine 98%
Supplier: Ambeed
|
Danger
|
A971825-250MG
A971825-1G
A971825-100MG
76994-068EA
35
USD
76994-068
77573-920
77886-939
4,4-dinitro-2,2-Bipyridine 98%
4,4-dinitro-2,2-Bipyridine
Precaución: for Research Use Only
Formula:
C₁₀H₆N₄O₄ |
MDL Number:
MFCD06657644 CAS Number: 18511-72-3 UN: 2926 ADR: 4.1,6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...