2-Methyl-3-(trifluoromethyl)benzonitrile 97%
Supplier: Ambeed
|
Danger
|
A970565-25G
A970565-5G
A970565-1G
77037-046EA
195
USD
77037-046
77037-048
77873-394
2-Methyl-3-(trifluoromethyl)benzonitrile 97%
2-Methyl-3-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₉H₆F₃N |
MDL Number:
MFCD01631593 CAS Number: 261952-02-7 UN: 3439 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...