2,4-Dichloropyridine 98%

Supplier: Ambeed
Warning

A970147-100G A970147-10G A970147-25G A970147-500G A970147-5G
76994-058EA 61.54 USD
76994-058 76994-060 76994-062 76994-064 76994-066
2,4-Dichloropyridine 98%
2,4-Dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
MDL Number: MFCD00955618
CAS Number: 26452-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR