Benzyl piperidin-3-ylcarbamate 98%

Supplier: Ambeed
Warning

A969353-1G A969353-250MG A969353-5G
76994-052EA 20.16 USD
76994-052 76994-054 76994-056
Benzyl piperidin-3-ylcarbamate 98%
Benzyl piperidin-3-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.29 g/mol
MDL Number: MFCD04115317
CAS Number: 31648-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR