3-Methyl-5-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A967530-100MG A967530-1G A967530-250MG A967530-5G
76994-044EA 40 USD
76994-044 76994-046 76994-048 76994-050
3-Methyl-5-nitrobenzaldehyde 98%
3-Methyl-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₃
MDL Number: MFCD18399644
CAS Number: 107757-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR