2-Amino-N-(2,2,2-trifluoroethyl)acetamide 95%

Supplier: Ambeed
Warning

A966803-100MG A966803-1G A966803-250MG A966803-5G
77037-030EA 5 USD
77037-030 77037-032 77037-034 77037-036
2-Amino-N-(2,2,2-trifluoroethyl)acetamide 95%
2-Amino-N-(2,2,2-trifluoroethyl)acetamide

Precaución: for Research Use Only

Formula: C₄H₇F₃N₂O
MDL Number: MFCD09944516
CAS Number: 359821-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR