tert-Butyl 4-bromo-3-fluorobenzoate 98%

Supplier: Ambeed
Warning

A966112-1G A966112-250MG
76994-030EA 43.5 USD
76994-030 76994-032
tert-Butyl 4-bromo-3-fluorobenzoate 98%
tert-Butyl 4-bromo-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrFO₂
MW: 275.11 g/mol
MDL Number: MFCD08703424
CAS Number: 1057961-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR