1-(4-Bromo-2-fluoro-6-hydroxyphenyl)ethanone 98%

Supplier: Ambeed
Warning

A965549-100MG A965549-250MG A965549-1G A965549-5G
77020-130EA 15 USD
77020-130 77020-132 77573-910 77614-680
1-(4-Bromo-2-fluoro-6-hydroxyphenyl)ethanone 98%
1-(4-Bromo-2-fluoro-6-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.03 g/mol
MDL Number: MFCD22570395
CAS Number: 1369594-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR