3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-(9-Phenyl-9H-carbazol-3-yl)phenyl)-[1,1'-biphenyl]-4-amine

A964045-1G A964045-250MG A964045-5G
77020-118EA 7 USD
77020-118 77020-120 77020-122
3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole 98%
3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole

Precaución: for Research Use Only

Formula: C₃₆H₂₆N₂
MDL Number: MFCD22571697
CAS Number: 1160294-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR