2-Chloro-N-((4-chlorophenyl)carbamoyl)benzamide 95%

Supplier: Ambeed
Warning

A963063-1G A963063-25G A963063-5G
77020-108EA 13 USD
77020-108 77020-110 77020-112
2-Chloro-N-((4-chlorophenyl)carbamoyl)benzamide 95%
2-Chloro-N-((4-chlorophenyl)carbamoyl)benzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₀Cl₂N₂O₂
MDL Number: MFCD08273838
CAS Number: 57160-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR