Benzyl 3,3-difluorocyclobutanecarboxylate 97%

Supplier: Ambeed
Warning

A962982-1G A962982-250MG A962982-5G
76994-024EA 124.13 USD
76994-024 76994-026 76994-028
Benzyl 3,3-difluorocyclobutanecarboxylate 97%
Benzyl 3,3-difluorocyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₂O₂
MW: 226.21 g/mol
MDL Number: MFCD18909480
CAS Number: 935273-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR