3-Fluoro-4-(trifluoromethoxy)benzaldehyde 97%

Supplier: Ambeed
Warning

A962691-1G A962691-250MG
76994-020EA 67.9 USD
76994-020 76994-022
3-Fluoro-4-(trifluoromethoxy)benzaldehyde 97%
3-Fluoro-4-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₄O₂
MDL Number: MFCD00236275
CAS Number: 473917-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR