3-Iodo-2-methylpyridin-4-amine 95%

Supplier: Ambeed
Warning

A960849-100MG A960849-250MG
77020-100EA 24 USD
77020-100 77020-102
3-Iodo-2-methylpyridin-4-amine 95%
3-Iodo-2-methylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₇IN₂
MDL Number: MFCD11656241
CAS Number: 849353-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR