Methyl 4-fluoro-2-methyl-5-nitrobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-(methoxycarbonyl)-4-methylnitrobenzene, 5-Fluoro-2-(methoxycarbonyl)-4-nitrotoluene

A960756-25G A960756-5G A960756-1G
76994-014EA 227.04 USD
76994-014 76994-016 77684-899
Methyl 4-fluoro-2-methyl-5-nitrobenzoate 95%
Methyl 4-fluoro-2-methyl-5-nitrobenzoate

Precaución: For research use only.

Formula: C₉H₈FNO₄
MW: 213.163 g/mol
MDL Number: MFCD14581697
CAS Number: 1163287-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR