3-bromo-N-methylpyridin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-N-methylpyridin-2-amine

A960571-10G A960571-1G A960571-250MG A960571-5G
76994-006EA 894.34 USD
76994-006 76994-008 76994-010 76994-012
3-bromo-N-methylpyridin-2-amine 98%
3-bromo-N-methylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.037 g/mol
MDL Number: MFCD07374349
CAS Number: 214977-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR