(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct 98%

Supplier: Ambeed
Warning

Synonyms: (2Rp)-1-[(1S)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene, (S)-(R)-JOSIPHOS, (2S)-1-[(1S)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene, Josiphos SL-J001-2, (S)-1-[(RP)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine

A959611-100MG A959611-250MG
76994-002EA 40.32 USD
76994-002 76994-004
(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct 98%
(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct

Precaución: for Research Use Only

Formula: C₃₆H₄₄FeP₂
MW: 594.53 g/mol
MDL Number: MFCD00800285
CAS Number: 162291-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR