3-Amino-4-phenylpyridine 97%

Supplier: Ambeed
Warning

A959426-100MG A959426-250MG A959426-1G
77020-096EA 31 USD
77020-096 77020-098 77891-720
3-Amino-4-phenylpyridine 97%
3-Amino-4-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂
MDL Number: MFCD04114255
CAS Number: 146140-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR