Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde 93%
Supplier: Ambeed
|
Warning
|
Synonyms:
Thieno[3,2-b]thiophene-2,5-dicarbaldehyde
A957954-100MG
A957954-1G
A957954-250MG
A957954-25G
A957954-5G
77020-086EA
28.65
USD
77020-086
77020-088
77020-090
77020-092
77020-094
Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde 93%
Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde
Precaución: for Research Use Only
Formula:
C₈H₄O₂S₂ MW: 196.24 g/mol |
MDL Number:
MFCD03931117 CAS Number: 37882-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...