tert-Butyl (4R,6R)-2-[6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 2-((4R,6R)-6-(2-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)ethyl)-2,2-dimethyl-1,3-dioxan-4-yl)acetate, Atorvastatin Acetonide tert-Butyl Ester
A957479-5G
A957479-1G
A957479-25G
A957479-100G
A957479-500G
77007-194EA
21
USD
77007-194
77603-352
77603-354
77674-416
77882-846
tert-Butyl (4R,6R)-2-[6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate 98%
tert-Butyl (4R,6R)-2-[6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Precaución: for Research Use Only
Formula:
C₄₀H₄₇FN₂O₅ |
MDL Number:
MFCD04039904 CAS Number: 125971-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...