4-Bromo-3-chloro-5-nitropyridine 95%

Supplier: Ambeed
Danger

A956192-10G A956192-1G A956192-250MG A956192-25G A956192-5G
77007-186EA 94 USD
77007-186 77007-188 77007-190 77007-192 77882-844
4-Bromo-3-chloro-5-nitropyridine 95%
4-Bromo-3-chloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClN₂O₂
MDL Number: MFCD11052835
CAS Number: 1072141-30-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR